Ethyl-3-Amino-2-methylbenzoate ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Varning

Synonyms: 3-Amino-o-toluic Acid Ethyl Ester, 3-Amino-2-methylbenzoic Acid Ethyl Ester, Ethyl 3-amino-2-methylbenzoate, Ethyl 3-Amino-o-toluate

A1574-25G A1574-5G
TCIAA1574-25GEA 3425 SEK
TCIAA1574-25G TCIAA1574-5G
Ethyl-3-Amino-2-methylbenzoate ≥98.0% (by GC, titration analysis)
Ethyl-3-Amino-2-methylbenzoate
Formel: C₁₀H₁₃NO₂
Kokpunkt: 150°C
MDL Number: MFCD00957246
CAS nummer: 57414-85-4

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR