2-(4-Aminophenyl)ethylamine ≥97.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: 4-(2-Aminoethyl)aniline, 1-Amino-2-(4-aminophenyl)ethane, 4-Aminophenethylamine

A1509-1G A1509-25G A1509-5G
TCIAA1509-1GEA 495 SEK
TCIAA1509-1G TCIAA1509-25G TCIAA1509-5G
2-(4-Aminophenyl)ethylamine ≥97.0% (by GC, titration analysis)
2-(4-Aminophenyl)ethylamine
Formel: C₈H₁₂N₂
MW: 136,20 g/mol
Kokpunkt: 142°C
MDL Number: MFCD00008194
CAS nummer: 13472-00-9
EINECS: 236-739-2
UN: 3259
ADR: 8,II

Order Now

Specifikation testresultat

Appearance White or Colorless to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR