1-Cyanovinyl acetate ≥97.0% stabilised

Leverantör: TCI EUROPE
Fara

Synonyms: 1-Acetoxy-1-cyanoethylene, Acetic Acid 1-Cyanovinyl Ester, 2-Acetoxyacrylonitrile

Stabilization: med 4-tert-Butylpyrokatekol

A1477-25G A1477-5G
TCIAA1477-25GEA 12650 SEK
TCIAA1477-25G TCIAA1477-5G
1-Cyanovinyl acetate ≥97.0% stabilised
1-Cyanovinyl acetate
Formel: C₅H₅NO₂
MW: 111,10 g/mol
Kokpunkt: 173°C
Flampunkt: 69°C
Storage Temperature: Kylskåp
MDL Number: MFCD00001868
CAS nummer: 3061-65-2
EINECS: 221-303-6
UN: 3276
ADR: 6.1,II

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR