4-Hydroxybutyl acrylate ≥97.0% stabilised

Leverantör: TCI EUROPE
Varning

Synonyms: Acrylic Acid 4-Hydroxybutyl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A1390-25G A1390-500G A1390-100G
TCIAA1390-25GEA 230 SEK
TCIAA1390-25G TCIAA1390-500G TCIAA1390-100G
4-Hydroxybutyl acrylate ≥97.0% stabilised
4-Hydroxybutyl acrylate
Formel: C₇H₁₂O₃
MW: 144,17 g/mol
Kokpunkt: 95°C
MDL Number: MFCD00010261
CAS nummer: 2478-10-6
EINECS: 219-606-3

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR