(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: (S)-(1-Ethylpyrrolidin-2-yl)methanamine

A1301-1G A1301-5G
TCIAA1301-1GEA 1225 SEK
TCIAA1301-1G TCIAA1301-5G
(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine ≥98.0% (by GC, titration analysis)
(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine
Formel: C₇H₁₆N₂
MW: 128,22 g/mol
Flampunkt: 57°C
MDL Number: MFCD00191371
CAS nummer: 22795-99-9
UN: 2733
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -94.0 to -97.0 deg (C=0.9, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR