2-(Dimethylamino)ethyl acrylate ≥98.0% stabilised
Leverantör: TCI
Fara
Stabilization: Stabilised with 4-methoxyphenol (MEHQ)
A1235-25G A1235-500G A1235-100G
TCIAA1235-25GEA 230 SEK
TCIAA1235-25G TCIAA1235-500G TCIAA1235-100G
2-(Dimethylamino)ethyl acrylate ≥98.0% stabilised
2-(Dimethylamino)ethyl acrylate
Formel: C₇H₁₃NO₂
Kokpunkt: 161°C
Flampunkt: 59°C
Storage Temperature: Kylskåp
MDL Number: MFCD00038233
CAS nummer: 2439-35-2
EINECS: 219-460-0
UN: 3302
ADR: 6.1,II
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %