4-Aminopyrimidine-5-carbonitrile ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

A1214-1G A1214-5G
TCIAA1214-1GEA 405 SEK
TCIAA1214-1G TCIAA1214-5G
4-Aminopyrimidine-5-carbonitrile ≥98.0% (by GC, titration analysis)
4-Aminopyrimidine-5-carbonitrile
Formel: C₅H₄N₄
MW: 120,11 g/mol
MDL Number: MFCD00054552
CAS nummer: 16357-69-0
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=611999e2be17ed6f8b23e1ecaeab2f2c5e11d36d&vid=8228e0b634c9d1d80879ae01e589ea6f72baccc8&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR