Cyclohexyl acrylate ≥98.0% stabilised
Leverantör: TCI
|
Varning
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
A1208-25ML
A1208-500ML
TCIAA1208-25MLEA
215
SEK
TCIAA1208-25ML
TCIAA1208-500ML
Cyclohexyl acrylate ≥98.0% stabilised
Cyclohexyl acrylate
Formel:
C₉H₁₄O₂ Kokpunkt: 184°C Flampunkt: 68°C |
MDL Number:
MFCD00046349 CAS nummer: 3066-71-5 EINECS: 221-319-3 UN: 3082 ADR: 9,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...