trans-4-Acetamidocyclohexanol ≥98.0%

Leverantör: TCI

A1162-25G A1162-5G
TCIAA1162-25GEA 1625 SEK
TCIAA1162-25G TCIAA1162-5G
trans-4-Acetamidocyclohexanol ≥98.0%
trans-4-Acetamidocyclohexanol
Formel: C₈H₁₅NO₂
MDL Number: MFCD00051422
CAS nummer: 27489-60-7
EINECS: 248-491-2

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 168.0 to 172.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR