4-Phenylbutylamine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-Amino-4-phenylbutane, 4-Phenylbutan-1-amine
A1123-25ML
A1123-5ML
TCIAA1123-25MLEA
1675
SEK
TCIAA1123-25ML
TCIAA1123-5ML
4-Phenylbutylamine ≥98.0% (by GC, titration analysis)
4-Phenylbutylamine
Formel:
C₁₀H₁₅N MW: 149,24 g/mol Kokpunkt: 144°C Flampunkt: 101°C |
MDL Number:
MFCD00008231 CAS nummer: 13214-66-9 EINECS: 236-186-7 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...