DL-ɑ-Amino-ε-caprolactam ≥98.0%
Leverantör: TCI
|
Varning
|
A1003-5G
A1003-1G
TCIAA1003-5GEA
2900
SEK
TCIAA1003-5G
TCIAA1003-1G
DL-ɑ-Amino-ε-caprolactam ≥98.0%
DL-alpha-Amino epsilon caprolactam
Formel:
C₆H₁₂N₂O Kokpunkt: 172°C |
MDL Number:
MFCD00064475 CAS nummer: 671-42-1 EINECS: 211-584-3 |
|
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 76.0 to 80.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...