3,5,5-Trimethylhexyl acetate ≥93.0%
Leverantör: TCI EUROPE
Synonyms:
Acetic Acid 3,5,5-Trimethylhexyl Ester
A0982-25ML
A0982-500ML
TCIAA0982-25MLEA
215
SEK
TCIAA0982-25ML
TCIAA0982-500ML
3,5,5-Trimethylhexyl acetate ≥93.0%
3,5,5-Trimethylhexyl acetate
Formel:
C₁₁H₂₂O₂ MW: 186,30 g/mol Kokpunkt: 209°C Flampunkt: 79°C |
MDL Number:
MFCD00026336 CAS nummer: 58430-94-7 EINECS: 261-245-9 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 93.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...