Oleyl acetate >60.0%
Leverantör: TCI EUROPE
Synonyms:
Acetic Acid Oleyl Ester
A0934-25ML
A0934-500ML
TCIAA0934-25MLEA
295
SEK
TCIAA0934-25ML
TCIAA0934-500ML
Oleyl acetate >60.0%
Oleyl acetate
Formel:
C₂₀H₃₈O₂ MW: 310,52 g/mol Kokpunkt: 186°C Flampunkt: 190°C |
MDL Number:
MFCD00056326 CAS nummer: 693-80-1 EINECS: 211-759-4 |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 60.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...