Acetrizoic acid ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Acetamido-2,4,6-triiodobenzoic Acid

A0858-1G A0858-5G
TCIAA0858-1GEA 685 SEK
TCIAA0858-1G TCIAA0858-5G
Acetrizoic acid ≥98.0%
Acetrizoic acid
Formel: C₉H₆I₃NO₃
MW: 556,86 g/mol
MDL Number: MFCD00016495
CAS nummer: 85-36-9
EINECS: 201-600-7

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR