ɑ-Amylcinnamaldehyde ≥90.0%
Leverantör: TCI
|
Varning
|
Synonyms:
Amylcinnamal, 2-Amyl-3-phenylacrylaldehyde, 2-Benzylideneheptanal, ɑ-Pentylcinnamaldehyde, 2-Amyl-3-phenyl-2-propenal, Jasminal
A0851-25ML
A0851-500ML
TCIAA0851-25MLEA
215
SEK
TCIAA0851-25ML
TCIAA0851-500ML
ɑ-Amylcinnamaldehyde ≥90.0%
alpha-Amylcinnamaldehyde
Formel:
C₁₄H₁₈O MW: 202.30 g/mol Kokpunkt: 175°C |
MDL Number:
MFCD00006988 CAS nummer: 122-40-7 EINECS: 204-541-5 UN: 3082 ADR: 9 III |
Specifikation testresultat
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...