4-Nitrophenyl anthranilate ≥98.0%

Leverantör: TCI
Varning

Synonyms: Anthranilic Acid 4-Nitrophenyl Ester, 2-Aminobenzoic Acid 4-Nitrophenyl Ester

A0842-25G A0842-5G
TCIAA0842-25GEA 5400 SEK
TCIAA0842-25G TCIAA0842-5G
4-Nitrophenyl anthranilate ≥98.0%
4-Nitrophenyl anthranilate
Formel: C₁₃H₁₀N₂O₄
MDL Number: MFCD00056116
CAS nummer: 19176-60-4

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 125.0 to 128.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR