6-Aminopenicillanic acid ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
(2S,5R,6R)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, (2S,5R,6R)-(+)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
A0800-25G
A0800-5G
TCIAA0800-25GEA
655
SEK
TCIAA0800-25G
TCIAA0800-5G
6-Aminopenicillanic acid ≥98.0%
6-Aminopenicillanic acid
Formel:
C₈H₁₂N₂O₃S MW: 216,26 g/mol |
MDL Number:
MFCD00005176 CAS nummer: 551-16-6 EINECS: 208-993-4 Merck Index: 14,00458 |
Specifikation testresultat
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Specific rotation [a]20/D | +270.0 to +280.0 deg (C=1.2, 0.1mol/L HCl) |
Solubility in 1mol/L HCl | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...