1-Adamantanecarbonyl chloride ≥97.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
1-Adamantanecarboxylic acid chloride
A0717-25G
A0717-5G
TCIAA0717-25GEA
1450
SEK
TCIAA0717-25G
TCIAA0717-5G
1-Adamantanecarbonyl chloride ≥97.0% (by GC, titration analysis)
1-Adamantanecarbonyl chloride
Formel:
C₁₁H₁₅ClO MW: 198,69 g/mol Kokpunkt: 136°C |
MDL Number:
MFCD00074724 CAS nummer: 2094-72-6 EINECS: 218-252-7 UN: 3261 ADR: 8,III |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Melting point | 51.0 to 54.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...