Ammelide ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Cyanuramide, Melanuric Acid, Dihydro-6-imino-1,3,5-triazine-2,4(1H,3H)-dione, 6-Amino-1,3,5-triazine-2,4-diol, Cyanuric Acid Monoamide
A0645-1G
A0645-5G
TCIAA0645-1GEA
1175
SEK
TCIAA0645-1G
TCIAA0645-5G
Ammelide ≥98.0%
Ammelide
Formel:
C₃H₄N₄O₂ MW: 128,09 g/mol |
MDL Number:
MFCD00049046 CAS nummer: 645-93-2 EINECS: 211-456-7 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot 1mol/L NaOH | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...