Arbutin ≥95.0%

Leverantör: TCI EUROPE

Synonyms: 4-Hydroxyphenyl beta-D-Glucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(4-hydroxyphenoxy)tetrahydro-2H-pyran-3,4,5-triol, Hydroquinone-beta-D-glucoside

A0522-25G A0522-5G
TCIAA0522-25GEA 1425 SEK
TCIAA0522-25G TCIAA0522-5G
Arbutin ≥95.0%
Arbutin
Formel: C₁₂H₁₆O₇
MW: 272,25 g/mol
MDL Number: MFCD00016915
CAS nummer: 497-76-7
EINECS: 207-850-3
Merck Index: 14,00773

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Melting point 197.0 to 202.0 deg-C
Specific rotation [a]20/D -63.0 to -68.0 deg (C=4, H2O) (calcd.on anh.substance)
Solubility in Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR