p-Anisaldehyd ≥99.0%
Leverantör: TCI
|
Varning
|
A0480-25ML
A0480-500ML
TCIAA0480-25MLEA
415
SEK
TCIAA0480-25ML
TCIAA0480-500ML
p-Anisaldehyd ≥99.0%
p-Anisaldehyd
Formel:
C₈H₈O₂ Kokpunkt: 248°C Flampunkt: 130°C |
MDL Number:
MFCD00003385 CAS nummer: 123-11-5 EINECS: 204-602-6 Merck Index: 14,00663 |
|
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...