(S)-2-(4-Aminobenzamido)pentanedioic acid ≥97.0%

Leverantör: TCI EUROPE

Synonyms: N-(4-Aminobenzoyl)-L-glutamic acid, N-(4-NH₂-Bz)-Glu-OH

A0442-1G A0442-5G
TCIAA0442-1GEA 805 SEK
TCIAA0442-1G TCIAA0442-5G
(S)-2-(4-Aminobenzamido)pentanedioic acid ≥97.0%
(S)-2-(4-Aminobenzamido)pentanedioic acid
Formel: C₁₂H₁₄N₂O₅
MW: 266,2500 g/mol
MDL Number: MFCD00042821
CAS nummer: 4271-30-1
EINECS: 224-261-7
Merck Index: 14,00426
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3699fef1bd99ebda074afcb56cf2e46d4258ebf&vid=d3699fef1bd99ebda074afcb56cf2e46d4258ebf&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) min. 97.0 %
Specific rotation [a]20/D -13.0 to -17.0 deg (C=2, 0.1mol/L HCl)
Solubility in hot Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR