5-Aminovaleric acid hydrochloride, low water content ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
5-Aminopentanoic acid hydrochloride
A0436-1G
A0436-5G
TCIAA0436-1GEA
390
SEK
TCIAA0436-1G
TCIAA0436-5G
5-Aminovaleric acid hydrochloride, low water content ≥98.0%
5-Aminovaleric acid hydrochloride
Formel:
H₂N(CH₂)₄CO₂H·HCl MW: 153,6073 g/mol |
MDL Number:
MFCD00012919 CAS nummer: 627-95-2 EINECS: 211-021-1 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 93.0 to 97.0 deg-C |
water | max. 200 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...