3-Amino-4-hydroxybenzenesulphonic acid hydrate ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-Hydroxymetanilic acid, 2-Aminophenol-4-sulphonic acid, 3-Amino-4-hydroxybenzenesulphonic acid
A0399-500G
A0399-25G
TCIAA0399-500GEA
1475
SEK
TCIAA0399-500G
TCIAA0399-25G
3-Amino-4-hydroxybenzenesulphonic acid hydrate ≥98.0%
3-Amino-4-hydroxybenzenesulphonic acid hydrate
Formel:
C₆H₇NO₄S MW: 189,19 g/mol |
MDL Number:
MFCD00007696 CAS nummer: 98-37-3 EINECS: 202-662-8 |
Specifikation testresultat
Appearance | Orange to Amber to Dark red powder to crystal |
Purity (Neutralization titration) | min. 98.0% (calcd.on anh.substance) |
Solubility in hot Water | almost transparency |
Water | max. 9.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...