2-Amino-4-nitrophenol sodium salt ≥80.0%
Leverantör: TCI
|
Varning
|
A0380-25G
A0380-500G
TCIAA0380-25GEA
215
SEK
TCIAA0380-25G
TCIAA0380-500G
2-Amino-4-nitrophenol sodium salt ≥80.0%
2-Amino-4-nitrophenol sodium salt
Formel:
C₆H₆N₂O₃ |
MDL Number:
MFCD00067428 CAS nummer: 61702-43-0 EINECS: 262-914-8 |
Specifikation testresultat
Appearance | Orange to Brown to Dark red powder to crystal |
Purity (Nonaqueous Titration) | min. 80.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...