2-Amino-4-methylthiazole hydrochloride ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
Synonyms:
4-Methyl-thiazol-2-ylamine hydrochloride
A0336-25G
A0336-5G
TCIAA0336-25GEA
1275
SEK
TCIAA0336-25G
TCIAA0336-5G
2-Amino-4-methylthiazole hydrochloride ≥98.0% (by HPLC, total nitrogen)
2-Amino-4-methylthiazole hydrochloride
Formel:
C₄H₆N₂S MW: 150,62 g/mol |
MDL Number:
MFCD00035214 CAS nummer: 6142-15-0 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 170.0 to 174.0 deg-C |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...