4-Amino-4'-methoxystilbene ≥97.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 4'-Methoxy-4-stilbenamine

A0326-1G A0326-5G
TCIAA0326-1GEA 1850 SEK
TCIAA0326-1G TCIAA0326-5G
4-Amino-4'-methoxystilbene ≥97.0% (by GC, titration analysis)
4-Amino-4'-methoxystilbene
Formel: C₁₅H₁₅NO
MW: 225,29 g/mol
MDL Number: MFCD00059856
CAS nummer: 7570-37-8

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 172.0 to 176.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR