4-Amino-3-hydroxybutyric acid ≥98.0%

Leverantör: TCI EUROPE
Varning

A0318-25G A0318-5G
TCIAA0318-25GEA 845 SEK
TCIAA0318-25G TCIAA0318-5G
4-Amino-3-hydroxybutyric acid ≥98.0%
4-Amino-3-hydroxybutyric acid
Formel: C₄H₉NO₃
MW: 119,12 g/mol
MDL Number: MFCD00008141
CAS nummer: 924-49-2
EINECS: 213-106-9
Merck Index: 14,00444

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR