2-Amino-3-hydroxyanthraquinone ≥98.0%
Leverantör: TCI EUROPE
A0315-25G
A0315-500G
A0315-5G
TCIAA0315-25GEA
925
SEK
TCIAA0315-25G
TCIAA0315-500G
TCIAA0315-5G
2-Amino-3-hydroxyanthraquinone ≥98.0%
2-Amino-3-hydroxyanthraquinone
Formel:
C₁₄H₉NO₃ MW: 239,23 g/mol |
MDL Number:
MFCD00059499 CAS nummer: 117-77-1 EINECS: 204-206-3 |
Specifikation testresultat
Appearance | Orange to Amber to Dark red powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in N,N-DMF | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...