4-Aminoantipyrine hydrochloride ≥98.0% (by HPLC, titration analysis) for biochemical research
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-AA.HCl, 4-Amino-1,5-dimethyl-2-phenyl-1H-pyrazol-3(2H)-one hydrochloride
A0257-25G
A0257-5G
TCIAA0257-25GEA
1200
SEK
TCIAA0257-25G
TCIAA0257-5G
4-Aminoantipyrine hydrochloride ≥98.0% (by HPLC, titration analysis) for biochemical research
4-Aminoantipyrine hydrochloride
Formel:
C₁₁H₁₃N₃O MW: 239,70 g/mol |
MDL Number:
MFCD00042012 CAS nummer: 22198-72-7 EINECS: 244-832-4 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Hydrogendioxide detection test (Peroxidase method) | to pass test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...