Diisodecyl adipate

Leverantör: TCI EUROPE

Synonyms: Adipic Acid Diisodecyl Ester

A0164-25ML A0164-500ML
TCIAA0164-25MLEA 215 SEK
TCIAA0164-25ML TCIAA0164-500ML
Diisodecyl adipate
Diisodecyl adipate
MW: 426,68 g/mol
Flampunkt: 229°C
MDL Number: MFCD00048392
CAS nummer: 27178-16-1
EINECS: 248-299-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28544aef7fd395b6d2c18ebe23b0ec5c9a3a21d3&vid=28544aef7fd395b6d2c18ebe23b0ec5c9a3a21d3&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR