N-Acetyl-L-tyrosine ethyl ester monohydrate ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE

Synonyms: Ac-Tyr-OEt·H2O

A0118-1G A0118-5G
TCIAA0118-1GEA 295 SEK
TCIAA0118-1G TCIAA0118-5G
N-Acetyl-L-tyrosine ethyl ester monohydrate ≥98.0% (by HPLC, total nitrogen)
N-Acetyl-L-tyrosine ethyl ester monohydrate
Formel: C₁₃H₁₉NO₅
MW: 269,297 g/mol
MDL Number: MFCD00149093
CAS nummer: 840-97-1
EINECS: 212-663-5

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Melting point 79.0 to 83.0 deg-C
Specific rotation [a]20/D +20.0 to +24.0 deg (C=1, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR