2-Acetamidofluorene ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
N-Fluoren-2-ylacetamide, N-(9H-Fluoren-2-yl)acetamide, N-2-Fluorenylacetamide, 2-AAF
A0076-25G
A0076-5G
TCIAA0076-25GEA
2450
SEK
TCIAA0076-25G
TCIAA0076-5G
2-Acetamidofluorene ≥98.0% (by HPLC, total nitrogen)
2-Acetamidofluorene
Formel:
C₁₅H₁₃NO MW: 223,28 g/mol |
MDL Number:
MFCD00001116 CAS nummer: 53-96-3 EINECS: 200-188-6 Merck Index: 14,04157 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 194.0 to 197.0 deg-C |
Solubility in hot EtOH | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...