tert-Butyl acetoacetate ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: tert-Butyl Acetoacetate
A11115.22 A11115.36
A11115.22EA 740 SEK
A11115.22 A11115.36
tert-Butyl acetoacetate ≥98%
tert-Butyl acetoacetate
Purity: 98+% CAS No.: 1694-31-1 Molecular Formula: C8H14O3 Formula Weight: 158.20 Flash Point: 76°(169°F) Refractive Index: 1.4200 Odour: Pleasant pH: 5.7 Melting Point: -38° Boiling Point: 71-72°/11mm Beilstein Reference: 1680303 MDL No.: MFCD00008811
Formel: C₈H₁₄O₃
MW: 158,2 g/mol
Kokpunkt: 71…72 °C (11 mmHg)
Smältpunkt: –38 °C
Densitet: 0,961
Flampunkt: 76 °C (169 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008811
CAS nummer: 1694-31-1
EINECS: 216-904-5