Malonnitril 99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Propandinitril
125271000 125275000 125270025
ACRO125271000EA 595 SEK
ACRO125271000 ACRO125275000 ACRO125270025
Malonnitril 99%
Malonnitril
Formel: C₃H₂N₂
MW: 66.06 g/mol
Kokpunkt: 220 °C (760 mmHg)
Smältpunkt: 30…34 °C
Densitet: 1.04 g/cm³
Flampunkt: 112 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001883
CAS nummer: 109-77-3
EINECS: 203-703-2
UN: 2647
ADR: 6.1 II
Merck Index: 13,05733
Order Now
Specifikation testresultat
Appearance White to yellow-brown Low melting crystalline mass
Infrared spectrum Conforms
Melting point 30 to 34 °C
GC ≥98.5 %