2-Metoxi-1-metyletylacetat ≥99% stabilised
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Propylene glycol 1-methyl ether 2-acetat, 2-(1-Methoxy)propyl acetate, 1-Methoxy-2-propyl acetate, Propylene glycol methyl ether acetat, 1,2-Propanediol monomethyl ether acetate, DOWANOL PMA
Stabilization: 50 ppm Stabilised med BHT/Ionol
L15459.AE L15459.AU L15459.0F
L15459.AEEA 725 SEK
L15459.AE L15459.AU L15459.0F
2-Metoxi-1-metyletylacetat ≥99% stabilised
2-Metoxi-1-metyletylacetat
Stabilized with 50 ppm BHT
Purity: 99% CAS No.: 108-65-6 Molecular Formula: C6H12O3 Formula Weight: 132.16 Flash Point: 43°(109°F) Refractive Index: 1.4020 Odour: Ester-like pH: 4 Melting Point: -67° Boiling Point: 145-146° Beilstein Reference: 1751656 UN No.: 3272 MDL No.: MFCD00038500
Formel: C₆H₁₂O₃
MW: 132.16 g/mol
Kokpunkt: 145…146 °C
Smältpunkt: –67 °C
Densitet: 0.968
Flampunkt: 43 °C (109 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00038500
CAS nummer: 108-65-6
EINECS: 203-603-9
UN: 3272
ADR: 3 III