Zirconylkloridk oktahydrat ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: Zirconium(IV) oxide chloride octahydrate, Zirconium dichloride oxide octahydrate, Zirconium(IV) oxychloride octahydrate
A12342.0E A12342.22 A12342.36
A12342.0EEA 25800 SEK
A12342.0E A12342.22 A12342.36
Zirconylkloridk oktahydrat ≥98%
Zirconylkloridk oktahydrat
Purity: 98% CAS No.: 13520-92-8 Molecular Formula: ZrOCl2.8H2O Formula Weight: 322.25 (178.13anhy) Odour: Acidic Melting Point: 150° dec. UN No.: 3260 MDL No.: MFCD00149898
Formel: Cl₂OZr·8H₂O
MW: 322,25 g/mol
Smältpunkt: 150 °C (dec.)
Densitet: 1,91
Storage Temperature: Omgivande
MDL Number: MFCD00149898
CAS nummer: 13520-92-8
EINECS: 231-717-9
UN: 3260
ADR: 8,II
Merck Index: 14,10184