L(+)-Lysine monohydrate ≥97%
Leverantör: Thermo Scientific
Synonyms: L-(+)-Lysine, Hydrate, (S)-(+)-2,6-Diaminohexanoic acid monohydrate, (S)-(+)-Lysine monohydrate, L-Lysine hydrate, (S)-(+)-2,6-Diaminocaproic acid monohydrate, H-Lys-OH monohydrate, K monohydrate, H-Lys-OH·H₂O, Lys monohydrate
A13040.14 A13040.22 A13040.36
A13040.14EA 1300 SEK
A13040.14 A13040.22 A13040.36
L(+)-Lysine monohydrate ≥97%
L(+)-Lysine monohydrate
Purity: 97% CAS No.: 39665-12-8 Molecular Formula: C6H14N2O2.H2O Formula Weight: 164.21 (146.19anhy) Melting Point: 210° dec. Beilstein Reference: 1722531 Merck Reference: 14,5636 Optical Rotation: +22.5° (c=2 in 6M HCl) MDL No.: MFCD00151035
Formel: H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O
MW: 164,2 g/mol
Smältpunkt: 210 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00151035
CAS nummer: 39665-12-8
EINECS: 200-294-2
Merck Index: 14,5636