Europium trinitrate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Scientific
Fara
Synonyms: Europium (III) nitrate hydrate
11296.06 11296.14
11296.06EA 5625 SEK
11296.06 11296.14
Europium trinitrate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Europium trinitrate hydrate
Purity: 99.99% CAS No.: 100587-95-9 Molecular Formula: Eu(NO3)3.xH2O (x≈6) Formula Weight: 337.97(anhy) Physical Form: Crystalline Aggregates Appearance: Colourless to pink Melting Point: 85° Merck Reference: 14,3904 UN No.: 1477 MDL No.: MFCD00149700
Formel: EuN₃O₉·xH₂O
MW: 337,98 g/mol (anhy)
Smältpunkt: 85 °C
Storage Temperature: Omgivande
MDL Number: MFCD00149698
CAS nummer: 100587-95-9
EINECS: 233-380-3
UN: 1477
ADR: 5.1,III
Merck Index: 14,3904