(S)-(-)-3-Aminopyrrolidine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

A1168-25G A1168-5G A1168-1G
TCIAA1168-25GEA 6725 SEK
TCIAA1168-25G TCIAA1168-5G TCIAA1168-1G
(S)-(-)-3-Aminopyrrolidine ≥98.0% (by GC, titration analysis)
(S)-(-)-3-Aminopyrrolidine
Formel: C₄H₁₀N₂
MW: 86,14 g/mol
Kokpunkt: 159 - 160 °C (1013 hPa)
Densitet: 0,98 g/cm³ (20 °C)
Flampunkt: 68°C
Storage Temperature: Omgivande
MDL Number: MFCD00143193
CAS nummer: 128345-57-3
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29c23f28743c01b248d75842adaa0e562236c6d2&vid=29c23f28743c01b248d75842adaa0e562236c6d2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 99.0 ee%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR