Ammoniumheptamolybdat tetrahydrat ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: Hexaammonium heptamolybdate tetrahydrate, Ammonium molybdate(VI) tetrahydrate, Ammonium molybdate tetrahydrate, Hexammonium heptamolybdate 4-hydrate, Molybdic acid ammonium salt tetrahydrate
A13766.0B A13766.18 A13766.30
A13766.0BEA 5300 SEK
A13766.0B A13766.18 A13766.30
Ammoniumheptamolybdat tetrahydrat ≥99%
Ammoniumheptamolybdat tetrahydrat
Purity: 99% CAS No.: 12054-85-2 Molecular Formula: (NH4)6Mo7O24.4H2O Formula Weight: 1235.86 (1163.79anhy) Melting Point: ca 190° dec. Merck Reference: 14,533 MDL No.: MFCD00167059
Formel: (NH₄)₆Mo₇O₂₄·4H₂O
MW: 1235,86 g/mol
Smältpunkt: ∼ 190 °C (dec.)
Densitet: 2,498
Storage Temperature: Omgivande
MDL Number: MFCD00167059
CAS nummer: 12054-85-2
EINECS: 234-722-4
Merck Index: 14,533