Gallium trinitrate hydrate ≥99.999%, Puratronic®
Leverantör: Thermo Fisher Scientific
Fara
11150.14 11150.22 11150.36 11150.06
11150.14EA 8225 SEK
11150.14 11150.22 11150.36 11150.06
Gallium trinitrate hydrate ≥99.999%, Puratronic®
Gallium trinitrate hydrate
Purity: 99.999% CAS No.: 69365-72-6 Molecular Formula: Ga(NO3)3.xH2O Formula Weight: 255.73(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Appearance: White Melting Point: 110° dec. Merck Reference: 14,4346 UN No.: 1477 MDL No.: MFCD00149723
Formel: GaN₃O₉·H₂O
MW: 273.75 g/mol
Smältpunkt: 110 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00149723
CAS nummer: 69365-72-6
EINECS: 236-815-5
UN: 1477
ADR: 5.1 III
Merck Index: 14,4346