Meglumine 99.0-101.0%, EMPROVE® API ChP, Ph. Eur., JP, USP suitable for use as an active pharmaceutical ingredient låga endotoxinhalter, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Synonyms: Meglumine, (-)-1-Deoxy-1-(methylamino)-D-glucitol, N-Methyl-D-glucamine, (-)-N-Methyl-D-glucamine, (2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol, (-)-1-Methylamino-1-deoxy-D-glucitol
1.06143.1000 1.06143.9025
1.06143.1000EA 7675 SEK
1.06143.1000 1.06143.9025
Meglumine 99.0-101.0%, EMPROVE® API ChP, Ph. Eur., JP, USP suitable for use as an active pharmaceutical ingredient låga endotoxinhalter, SAFC®
D-(-)-N-Methylglucamine
Formel: H₃CNHCH₂(CH(OH))₄CH₂OH
Kokpunkt: 210 °C (1013 hPa) (dec.)
Smältpunkt: 128…131 °C
Densitet: 300 kg/m3
Storage Temperature: Omgivande
MDL Number: MFCD00004707
CAS nummer: 6284-40-8
EINECS: 228-506-9
Merck Index: 12,06154