L (-)-Cystin Reag. Ph. Eur. pure, pharma grade
Leverantör: PanReac AppliChem
Synonyms: (R)-(-)-Cystine, (2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid), (H-Cys-OH)2
A1703.0100 A1703.1000
A1703.0100EA 1350 SEK
A1703.0100 A1703.1000
L (-)-Cystin Reag. Ph. Eur. pure, pharma grade
L (-)-Cystin
Formel: HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
MW: 240,3 g/mol
Kokpunkt: 428 °C (1013 hPa)
Smältpunkt: 261…262 °C
Densitet: 1,677 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00064228
CAS nummer: 56-89-3
EINECS: 200-296-3
Merck Index: 13,02811