L (+)-Serin, EMPROVE® EXPERT Ph. Eur., USP suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
1.07647.1000 1.07647.9010
1.07647.1000EA 20350 SEK
1.07647.1000 1.07647.9010
L (+)-Serin, EMPROVE® EXPERT Ph. Eur., USP suitable for use as excipient, SAFC®
L (+)-Serin
Formel: C₃H₇NO₃
Kokpunkt: 259 °C (1013 hPa)
Smältpunkt: 215…225 °C
Densitet: 1,6 g/cm³ (22 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00064224
CAS nummer: 56-45-1
EINECS: 200-274-3
Merck Index: 14,00846