Dodecamolybdofosforsyra hydrate ACS
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Molybdophosphoric acid hydrate, 12-Molybdophosphoric acid hydrate, Phosphomolybdic acid hydrate
56166.22 56166.06 56166.14 56166.36
56166.22EA 3425 SEK
56166.22 56166.06 56166.14 56166.36
Dodecamolybdofosforsyra hydrate ACS
Dodecamolybdofosforsyra hydrate
CAS No.: 51429-74-4 Molecular Formula: H3PO4.12MoO3.xH2O Formula Weight: 1825.25(anhy) Physical Form: Granular Density: 3.15 Sensitivity: Moisture Sensitive Appearance: Yellow Melting Point: 78-90° Merck Reference: 14,7342 UN No.: 3085 MDL No.: MFCD00149913
Formel: H₃[P(Mo₃O₁₀)₄]·nH₂O
MW: 1843.27 g/mol
Smältpunkt: 78…90 °C
Densitet: 3.15
Storage Temperature: Omgivande
MDL Number: MFCD00149913
CAS nummer: 51429-74-4
EINECS: 234-713-5
UN: 3085
ADR: 5.1 III
Merck Index: 14,7342