Gallussyra monohydrat ≥99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 3,4,5-Trihydroxybenzoic acid monohydrate, 3,4,5-Trihydroxybenzoic acid hydrate
A16303.30 A16303.0B
A16303.30EA 1100 SEK
A16303.30 A16303.0B
Gallussyra monohydrat ≥99%
Gallussyra monohydrat
Purity: 99% CAS No.: 5995-86-8 Molecular Formula: C7H6O5.H2O Formula Weight: 188.14 (170.12anhy) Melting Point: 250° dec. Merck Reference: 14,4345 MDL No.: MFCD00149098
Formel: C₇H₆O₅·1H₂O
MW: 188.14 g/mol
Smältpunkt: 250 °C (dec.)
Flampunkt: 250 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00149098
CAS nummer: 5995-86-8
EINECS: 205-749-9