Gallussyra monohydrat ≥98% ACS
Leverantör: Thermo Scientific
Varning
Synonyms: 3,4,5-Trihydroxybenzoic acid monohydrate, 3,4,5-Trihydroxybenzoic acid hydrate
36417.22 36417.36
36417.22EA 1650 SEK
36417.22 36417.36
Gallussyra monohydrat ≥98% ACS
Gallic acid monohydrate
Purity: 98+% CAS No.: 5995-86-8 Molecular Formula: C7H6O5.H2O Formula Weight: 188.14 (170.12anhy) Physical Form: Powder Sensitivity: Light Sensitive Appearance: White to cream Melting Point: 250° dec. Merck Reference: 14,4345 MDL No.: MFCD00149098
Formel: C₇H₆O₅·1H₂O
MW: 188,14 g/mol
Smältpunkt: 250 °C (dec.)
Flampunkt: 250 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00149098
CAS nummer: 5995-86-8
EINECS: 205-749-9
Merck Index: 14,4345