L (+)-Lysin monohydroklorid, EMPROVE® EXPERT Ph. Eur., BP, USP suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Synonyms: Lysine Hydrochloride, (S)-(+)-2,6-Diaminohexanoic acid monohydrochloride, L-Lysine monohydrochloride, H-Lys-OH·HCl, L-Lysine monohydrocholoride, L-Lysine Hydrochloride, (S)-(+)-Lysine monohydrochloride, (S)-(+)-2,6-Diaminocaproic acid monohydrochloride, H-Lys-OH.HCl
1.05701.1000 1.05701.5000 1.05701.9025
1.05701.1000EA 5025 SEK
1.05701.1000 1.05701.5000 1.05701.9025
L (+)-Lysin monohydroklorid, EMPROVE® EXPERT Ph. Eur., BP, USP suitable for use as excipient, SAFC®
L (+)-Lysin monohydroklorid
Formel: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
MW: 182,65 g/mol
Smältpunkt: 263…264 °C
Storage Temperature: Omgivande
MDL Number: MFCD00064564
CAS nummer: 657-27-2
EINECS: 211-519-9