1-Pentanesulphonic acid sodium salt monohydrate för HPLC
Leverantör: Thermo Fisher Scientific
Synonyms: Sodium 1-pentanesulphonate monohydrate
15714.06 15714.14 15714.22
15714.06EA 1425 SEK
15714.06 15714.14 15714.22
1-Pentanesulphonic acid sodium salt monohydrate för HPLC
1-Pentanesulphonic acid sodium salt monohydrate
CAS No.: 207605-40-1 Molecular Formula: C5H11NaO3S.H2O Formula Weight: 192.21 (174.19anhy) Physical Form: Crystalline powder Appearance: White Beilstein Reference: 3725147
Ion pair reagent
Formel: C₅H₁₁NaO₃S·1H₂O
MW: 192.21 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00007541
CAS nummer: 207605-40-1
EINECS: 245-208-4